SP Bel-Art Captor™ Pen Holder, Bel-Art Products, a part of SP
# de catalogue 47751-018
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Description:Pen holder
- Cat. no.:47751-018
- W×D×H:25.4×25.4×50.8 mm (1×1×2")
- Supplier No.:133730000
Spécifications
A propos de cet article
Designed to keep a marking pen easily accessible in a lab environment, this device allows quick use when only one hand is free.
- Molded polycarbonate construction
- Attach to any flat surface with adhesive backing
- Pen used must feature pocket clip
Holder can be attached to any flat surface with its adhesive-backed, press-fit Dual Lock™ reclosable fastener. Molded from tough, black polycarbonate, the holder is ideal for use on or around an autoclave, incubator, centrifuge, storeroom, freezer, or lab bench. The pen used must have a cap with a pocket clip for attachment to the pen holder well.
Customers Who Bought This Also Bought