VWR® Contour™ Suspended Cabinets
Fournisseur: VWR International
New Product
Certificats
A propos de cet article
Powder-coated, steel, suspended cabinets mount directly to VWR® Lab tables.
- Cabinets meet SEFA standards
- Excellent chemical resistance
- Soft close, positive catch doors
Suspended cabinets are available in a snow white or light neutral finish, with a variety of door and drawer configurations.
Ordering information: Mounting bar not included.
Customers Who Bought This Also Bought