Nunc™ Cell Factory™, Large Scale Production, Sterile, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Cell factory units maximize available growing area with low risk of contamination and easy handling
- Polystyrene construction
- Easy to customize and close
- Large-scale production
Molded from high-quality polystyrene with the Nunclon™Δ tissue culture treated surface. Each cell factory is a sealed stack of chambers with common vent and fill ports. The 40-chamber factory can be used in place of 30 roller bottles. Openings connecting the chambers cause media to fill evenly for consistent growth conditions. Vents can be capped or fitted with bacterial air vents.
They also offer considerable space reduction.
Ordering information: Each cell factory is supplied with a Nunclon™Δ certificate for efficient cell attachment, growth, and quality. The starter kit (62407-103) includes an HDPE connector, Tyvek® cell factory caps, sealing caps, tubing, clamp, and Pall® Life Sciences bacterial air vent.
Customers Who Bought This Also Bought