Nalgene® Marking Pens, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Permanent black ink, pocket clip felt-tip pens for writing on glass and plastic surfaces
- Markings will not be affected by autoclaving
Customers Who Bought This Also Bought