Serum Bottle, Borosilicate Glass, Aluminum Seal, KIMBLE®, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Kimble serum bottle is constructed from a unique borosilicate glass that offers the highest known resistance to chemical attack.
- GPI neck finish: 20A
Ideal for packaging, storage, and application of injectable and parenteral solution requiring the utmost purity. Bottle also offers a high resistance to thermal and mechanical shock.
Certifications: Meets or exceeds USP Specifications for Type I Glass Containers.
Ordering information: Bottle is not supplied with a closure.
Customers Who Bought This Also Bought