Embedding Cassettes, Strip Holed, Two-Piece
Fournisseur: FOUR E’S INDUSTRIAL GROUP INC.
New Product
Certificats
A propos de cet article
POM embedding cassettes ensure precise tissue processing with minimized deformation.
- POM material minimizes wax block deformation
- Even and uniform printing surfaces
- Front-hinged design allows easy cover removal
- 45° angled writing surface for convenient labeling
- For use with laser printers
Crafted from POM material, these embedding cassettes offer exceptional durability and minimize wax block deformation during tissue processing. Printing and labeling surfaces are perfectly flat, enabling high-quality labeling with text and data matrix codes. These cassettes offer a front-hinged design (also known as reverse hinged). The detachable cover can be easily removed and re-attached as many times as desired.
Packaging: Sleeved packaging: 90 per sleeve, 20 sleeves per case. Bulk packaging: 500 per bag, 6 bags per case.
Customers Who Bought This Also Bought