KIMAX® Reservoir Micro Burets, Class A, Straight Bore PTFE Stopcocks, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Precision bore buret with a straight bore PTFE stopcock designed for micro applications
Manufactured according to ASTM E287 and calibrated to ASTM E694.
The permanently attached 70mL reservoir, separate PTFE stopcock and side fill tube allow quick refilling. Reservoir fitted with a medium length [ST] 14/20 stopper. Durable white ceramic scale.
Ordering information: Shipped complete with buret, buret stopcock, side fill stopcock and reservoir stopper in NSTA approved packaging.
Customers Who Bought This Also Bought