Specifications
- Temperature range:–100...300 °C (meter)
- Description:RTD Platinum Thermometer
- Temperature resolution:0.01 °C (meter)
- Temperature accuracy:± (0.1% + 0.2 °C) below 200°C, ± (0.15% + 0.5 °C) otherwise (meter)
- Probe:1, Platinum RTD stainless-steel, 5.9" probe, 0.1" Ø
- Probe cable length:43"
- W×D×H:6.3×2.04×0.86”
- Weight:7.05 oz.
- Cat. no.:23609-228
Specifications
About this item
State-of-the-art aerospace technology delivers high platinum performance.
- Traceable®
- Celsius units
- Rigid housing design and the real-time response RTD probe
- Fast data updating in 1 second time interval
- Complete set IP67 waterproof and washable in washing machine
- Follow HACCP principle to check food safety
- Green/Red dual color backlight to indicate temperature hazards
- Countdown timer function included
- Hold function to freeze current reading
- One touch to display temperature change from baseline
- Non-sleep mode function available
- Big LCD for easy reading
- Max/Min reading display
- Temperature high alarm threshold programmable
High-precision thermometer is geared specifically for measuring temperatures in soil, foods, ovens, cuvettes, gas systems, water baths, wastewater, incubators, chemical solutions, petroleum products, and extremely cold conditions.
Innovative 0.01° resolution digital thermometer developed specifically for lab and plant use is the finest-performing RTD Platinum Thermometer ever designed. Accuracy of ± (0.1% + 0.2°C) below 200°C, ± (0.15% + 0.5°C) otherwise makes it ideal for experimental requirement. Ease of use makes it perfect for routine quality-control needs. One-button operation allows everyone in the lab to report identical readings and eliminates operator technique errors. State-of-the-art, fourth-generation aerospace engineering incorporates the most modern and sophisticated electronics in temperature measurement to achieve utmost in precision, stability, and dependability.