KIMAX® Acidity Test, Automatic Zero, Kimble Chase
# de catalogue 48726-049
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Acidity Test Kit
- Cat. no.:48726-049
- Supplier No.:620F-1
Spécifications
A propos de cet article
Used to determine the acidity of milk, cream, cheese, whey, and other dairy products
- Stopcock Size: 2
The supplied buret has a PTFE stopcock plug and is graduated with a durable white ceramic enamel scale from 0 to 1%, giving readings directly in percentage of acidity. The stirring rod, a glass tube closed at both ends, contains a paper slip with two bands of different shades of pink to assist in determining when the right amount of neutralizer has been added. Indicator is 1% phenolphthalein in 70% denatured ethanol (neutralizer is 0.1N sodium hydroxide [NaOH]—fully diluted). Volume: 1% (10mL) with subdivisions at 0.01% (0.1mL).
Ordering information: Kit consists of one each of the buret with PTFE stopcock plug, filling tube assembly (glass tubes and rubber stoppers), 100mL beaker, rubber bulb, reservoir bottle, stirring rod, clamp, and 9mL pipet.
Customers Who Bought This Also Bought