VWR® Alkali Burets, Automatic Self-Zeroing, Class A, Glass Stopcock
# de catalogue 17582-002
Fournisseur: VWR International
Spécifications
- Capacity:50 ml
- Graduation Interval:0.10 ml
- ST Joint No.:29/42
- Cat. no.:17582-002
- Reservoir Capacity:2000 ml
Spécifications
A propos de cet article
Borosilicate glass buret assembly for use with alkali solutions.
- Ready-to-Use alumina-filled drying tubes
Fill and inlet openings are fitted with drying tubes filled with activated alumina
Meets or exceeds Class A specifications for burets.
Glass stopcock and reservoir bottle have close-fitting standard taper joints. Durable white scale. Drying tubes may be regenerated by heating.
Ordering information: Shipped complete with buret, bottle, bottle adapter assembly, glass check valve, squeeze bulb, stopcock, and two alumina filled drying tubes.
Customers Who Bought This Also Bought