Autoclavable Shelving and Accessories, Metro™
Fournisseur: Metropolitan Wire
Certificats
A propos de cet article
Preconfigured shelving is ideal for autoclaving cages and other equipment
Autoclavable covers are rated for approximately 70 cycles. Casters have a load rating of 136 kg (300 lbs.) and are temperature rated to 149 °C (300 °F).
Ordering information: Cage racks come with aluminum split sleeves and swedged posts. Autoclavable casters must be ordered separately.
Customers Who Bought This Also Bought