Acid Burets, Auto Self-Zeroing, Glass Stopcock, AMK Glass
Fournisseur: AMK GLASS INC
Certificats
A propos de cet article
Borosilicate glass buret assembly for use with acids
The acid-resistant polypropylene adapter fits a standard 5-pint screw top reagent bottle.
Ordering information: Buret assembly includes: buret, rubber bulb, and bottle adapter. Bottle not included.
Customers Who Bought This Also Bought