Stainless Steel Frame Tool Cart and Stainless Steel LocBoard®
# de catalogue 77269-704
Fournisseur: Triton Products
New Product
Spécifications
- Width:21.25"
- Color:Silver
- Description:Stainless steel frame tool cart and stainless steel LocBoard®
- Height:39.25"
- Material:Stainless steel
- Depth:36.75"
Spécifications
A propos de cet article
Triton Mobile Tool Carts puts your tools right where you need them and keeps you organized. Choose from a variety of pegboards to meet your needs.
- Pull handle and durable neoprene casters (2 fixed and 2 swivel locking) make it easy to transport the carts across flat surfaces
- Narrow base allows the user to transport tools around tight corners and through narrow door openings
- Sturdy stainless steel bottom tray provides 4.9 sq. ft. of additional storage on the bottom with a shelf at the top of the cart
- 18 sq. ft. of storage surface
- 800 lbs. load rating
Stainless Steel LocBoard® Mobile Tool Carts offer the most secure and durable way to transport tools, supplies and parts anywhere within a facility. Stainless LocHook® 60000 series keep tools and items safe and securely in place. Dual-sided storage allows users to organize, manage and store precision tools, hand tools and equipment without clutter.
Ordering information: Limited lifetime warranty.
Customers Who Bought This Also Bought