Zinc Plated Steel Hook Assortment for DuraBoard® or ¹/₈" and ¹/₄" Pegboard, 36-Piece
# de catalogue 77269-366
Fournisseur: Triton Products
New Product
Spécifications
- Color:Silver
- Description:Zinc Plated steel hook Assortment for DuraBoard® or ¹/₈" and ¹/₄" Pegboard, 36-Piece
Spécifications
A propos de cet article
DuraHook® assortments save you time, money and space. Many of the assortments contain everything you’ll need to tackle your storage challenge.
- Unique DuraHook styles along can handle any pegboard storage requirement
- Innovative double-locking system locks the hook in place with a locking tab that mounts to the pegboard from the front
- Use on any ¹/₈" or ¹/₄" pegboard (excluding steel)
- Hooks mount vertically and horizontally in 4 distinct positions allowing the use of multiple hooks for large, heavy and odd-shaped items
- Fixtures install from the front, so no need to remove the board for installation or repositioning of DuraHooks
The DuraHook® 76936 is a 36 piece hook assortment kit. It contains the first double locking pegboard hooks with new and useful styles including spring clips, Extended Spring Clip and tool holder. Hook styles are: (6) 1" Single Rod 30 Degree Bend; (1) 6" Single Rod 30 Degree Bend; (6) 2¹/₂" Single Rod 30 Degree Bend; (4) Pliers/Wrench Holder; (5) Extended Spring Clip; (4) Standard Spring Clips; (5) Single Ring Tool Holder; (5) Curved Hook.
Ordering information: Limited lifetime warranty.
Customers Who Bought This Also Bought