ContactUsLinkComponent
Curved Stainless Steel Pegboard Hook for Stainless Steel LocBoard®
Catalogue # 77269-308
Fournisseur:  Triton Products
undefined
Curved Stainless Steel Pegboard Hook for Stainless Steel LocBoard®
Catalogue # 77269-308
Fournisseur:  Triton Products
Supplier Number:  65200
Spécifications
  • Color:
    Silver
  • Description:
    Curved stainless steel pegboard hook for stainless steel LocBoard®, 2.25" length, 2" Int. Ø, 3 per pack
  • Cat. no.:
    77269-308
Spécifications
À propos de cet article
The Stainless Steel LocHook® is ideal for food, medical, healthcare, restaurant, marine and commercial applications as well as any clean room or harsh environment that must meet stringent requirements.
  • Use in pairs to hold power tools including drills, caulk guns and many other long items
  • Easily repositioned to accommodate changing storage needs with no damage to the stainless steel LocBoard
  • Won't rust or corrode in any environment, even ones with extreme temperatures
  • Use multiple hooks for large, heavy and odd-shaped items
Having clean, easily accessible and versatile storage and organizational solutions is an issue that every business encounters. The food and healthcare industries must meet stringent requirements and the stainless steel pegboard storage from Triton Products provides the ideal answer. Two innovative best-selling products, the LocBoard and the LocHook, are now available in stainless steel. They provide a commercial grade heavy-duty pegboard and hooks system. The LocHook is an innovative pegboard hook which can be mounted in any direction and is rugged enough to support just about anything. Together, the LocBoard and LocHook will provide the perfect stainless steel pegboard solution. Imagine a clean, well-kept kitchen with every pan, ladle, knife and utensil conveniently hanging and within grasp at all times. Imagine every medical supply in a hospital being easily accessible at a moment's notice. That is the power of the stainless steel pegboard system from Triton Products.
Ordering information: Limited lifetime warranty.