Pre-Cleaned Glass Storage Jugs, Amber, Environmental Express
Supplier: Environmental Express
Certificates
About this item
All containers meet one of two levels of pre-cleaning.
Amber glass jugs include polypropylene closure and PTFE liner.
Level III indicates that containers have not received an EPA washing treatment. Level I indicates that each case of containers has been custody sealed, lot number labeled for traceability, and a Certificate of Analysis for EPA quality assurance treatment.
Customers Who Bought This Also Bought