KIMAX® Powder Funnels, Kimble Chase, DWK Life Sciencese
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Funnels feature short, wide stems and fire-polished tops and stems.
These powder funnels are made of glass.
Certifications: Designed from ASTM specification E1095, Type 4 requirements.
Spécifications
- Catalog No:
- 89001-416
- 89001-422
- 89001-424
- 89001-418
- 89001-420
- Stem length:
- 35 mm
- 35 mm
- 35 mm
- 35 mm
- 35 mm