Nalgene® Large Square Bottle, Polypropylene, Wide Mouth, Thermo Scientific
# de catalogue 16130-005
Fournisseur: Thermo Fisher Scientific
Spécifications
- Capacity:4 L (1 gal.)
- Cat. no.:16130-005
- Supplier No.:2122-0010
Spécifications
A propos de cet article
Space-saving bottle features convenient, molded-in handgrips.
- Excellent for storing and sterilizing contaminated slides and other small lab items
- Not leakproof
- Autoclavable
- Cap size: 100-415
Note: Before autoclaving, set cap on top of the bottle without engaging threads.
Ordering information: Supplied with polypropylene screw caps assembled on bottles.
Customers Who Bought This Also Bought