VWR® Large Wide Mouth Sample Bottle, HDPE
Supplier: VWR International
Certificates
About this item
Leakproof bottles are ideal for dry chemicals, specimen collecting, and water sampling.
- Leakproof
- Easy-to-grip integrated handle
- HDPE construction
They can be stored in a freezer to −100 °C (−148 °F). The 4 L (1 gal.) bottles feature an integrated handle for better grip, greater safety, and pouring convenience.
Ordering information: Size 100-415 polypropylene screw caps included.
Frequently Bought Together