Nalgene® Large Sample Bottles, High-Density Polyethylene, Wide Mouth, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
Widely used for collecting specimens and water sampling.
- Leakproof
- With polypropylene screw closure
Also excellent for use with dry chemicals. Mouth opening: 89 mm (3¹/₂").
Customers Who Bought This Also Bought