MiraWIPE® Wipers, Foamtec International
Fournisseur: Foamtec International
Certificats
A propos de cet article
Constructed from continuous filament microfiber polyester, woven wipers feature superior absorbency and contamination pick-up when compared to traditional knitted polyester wipers.
Woven, abrasion-resistant fabric minimizes particle generation from wiper face and edge.
Ordering information: Cleanroom laundered and packaged. Wipers are available in configurations to meet ISO Class 3 to 6 requirements; contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought