SP Wilmad-LabGlass Graduated Cylinders, with Detachable Plastic Base and Guard, TD, SP Industries
Fournisseur: Avantor
Certificats
A propos de cet article
These borosilicate glass graduated cylinders are calibrated "to deliver" (TD).
- Cylinders feature a single metric scale
- Supplied with glass cylinder, detachable plastic base, and plastic collar
- See LG-5517A and LG-5518 for replacement parts
Spécifications
- Catalog No:
- 14212-512
- Form:
- Short