KIMAX® Educational Grade Graduated Cylinders, Plastic Base, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Educational grade cylinders are made of sturdy borosilicate glass, with a reinforced top bead, pour spout, and hexagonal plastic base
Designed from ASTM Specification E1272, Style 1, Class B requirements.
Single metric scale is durable blue ceramic enamel. The 10mL cylinder has an enlarged funnel-shaped top for ease of filling. The 10mL and 25mL cylinders are comparatively short to provide increased stability. Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Specifications
- Catalog No:
- 89000-754
- 89001-158
- 89001-166
- 89001-168
- 89001-164
- 89001-170
- 89001-160
- 89001-162
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short