PYREX® Filtering Flasks, Heavy Wall, Replaceable Tubulation, Corning
Fournisseur: Corning
Certificats
A propos de cet article
PYREX® borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These filtering flask have bent, poly tubulation in a grommet which acts as a shock absorber. Graduated to show approximate capacity.
Spécifications
- Catalog No:
- 89090-734
- 29416-328
- 29416-306
- 89091-200
- 89091-290
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey