LPG Corrosion Test Cylinder, Koehler
# de catalogue 52933-560
Fournisseur: Koehler
Small Business Enterprise
Spécifications
- Description:LPG Corrosion Test Cylinder
- Cat. no.:52933-560
- Supplier No.:K40000
Spécifications
A propos de cet article
Stainless steel cylinder with 1/4" needle valves for purging and admitting LPG samples
Conforms to ASTM D1838 and related specifications.
Dip tube with hook suspends copper strip in sample. Knurled, threaded cap with O-ring gasket hand tightens to a positive seal. Withstands hydrostatic test pressure of 1000psig (6895kPa).
Customers Who Bought This Also Bought