KIMAX® Graduated Cylinders, Class B, To Contain, Kimble Chase
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Certificats
A propos de cet article
Designed from ASTM E1272, Style 1, Class B requirements. The 10mL size has funnel top and sizes 25mL and larger have SAFE-GARD® bumpers.
- Calibrated to contain at 20°C
- Single metric scale is durable white ceramic enamel
- Hexagonal base provides stability
Spécifications
- Catalog No:
- 89001-104
- 89001-106
- 89001-100
- 89001-096
- 89001-102
- 89001-094
- 89001-098
- 89001-108
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short