KIMAX® Graduated Cylinders, Class B, To Deliver, with Red Stripe, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Cylinders are made of sturdy borosilicate glass
Designed from ASTM Specification E1272, Style 1, Class B requirements.
A clear, reverse image in the single metric scale is highlighted against a ruby red stripe. The 10mL cylinder has an enlarged funnel-shaped top for ease of filling. Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Spécifications
- Catalog No:
- 89001-126
- 89001-140
- 89001-136
- 89001-138
- 89001-132
- 89001-134
- 89001-128
- 89001-130
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall