Men's Flame-Resistant/Chemical Splash Protection Lab Coats
Fournisseur: Bulwark Protection
New Product
Certificats
A propos de cet article
This environment of ingenuity and innovation demands reliable lab PPE that provides the ultimate in protection and comfort, for all-day wear. That’s why Bulwark CP offers breakthrough safety wear designed specifically for the lab.
- Black lay down collar for easy recognition as a CP lab coat
- Concealed snap front closure
- Splash-resistant pass-throughs
- FR only knit cuffs, safer than typical open cuff
- Double needle top-stitched chest pocket with pencil stall
- Two double needle top-stitched lower pockets with flaps
- Back vent for ease of walking and sitting
Regular clothing or lab coats simply won’t keep lab workers protected in the case of a thermal incident or inadvertent chemical splash, but these single and multi-hazard lab coats help provide protection that fits well and is easy to wear.
Certifications: NFPA 2112 compliant.
Spécifications
- Catalog No:
- 77322-572
- 77322-576
- 77322-570
- 77322-578
- 77322-582
- 77322-580
- 77322-574
- 77322-584
- Garment application:
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Fire protection, Chemical protection
- Gender:
- For men
- For men
- For men
- For men
- For men
- For men
- For men
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps