Jar, Polypropylene, Natural, Wide Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Natural polypropylene jars are translucent and durable.
- Polypropylene jars are autoclavable and typically have a temperature tolerance between 32 to 275 °F.
Jars are ideal for storing and collecting specimens and lab supplies. The wide neck and straight-sided design facilitates easy recovery of contents. Polypropylene offers greater tolerance to high temperatures than HDPE, as well as better contact clarity.
Ordering information: Jars 89093-116, -118, -166, -168, and -170 are supplied vacuumed and ionized.
Customers Who Bought This Also Bought