Chemware® Bottles, Teflon® PFA, Wide Mouth, Saint-Gobain
Fournisseur: Avantor
Certificats
A propos de cet article
Good for trace analysis. Leakproof, unbreakable PFA bottles for storage of virtually all corrosive chemicals including HF, nitric and perchloric acids.
- Bottles withstand temperatures from 500 °F (260 °C) to the cryogenic range without becoming brittle
- Chemically inert, non-stick surface is easy to clean
- PFA cap
Customers Who Bought This Also Bought