Leak-Resistant Bottles, High-Density Polyethylene, Wide Mouth, Wheaton®, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Durable bottles are ideal for general laboratory use and field sample analysis.
- Wide mouth design facilitates easy filling and pouring
- Designed with a special pour lip to facilitate safe, clean dispensing
- Double-seal closure for leak-resistance
Ordering information: Supplied in convenience-packed cartons with polypropylene caps attached.
Customers Who Bought This Also Bought