About this item
This tough yet flexible film is made of low-density, puncture-resistant polyethylene Ultraclean Film™ for applications requiring Level II (poly) and III (antistatic poly) packaging cleanliness.
- Thickness: 6 mil
- Tubing can be easily heat sealed
- Chemically resistant to most solvents
- Low water vapor transmission rate
Pink antistatic poly bags and tubing feature good electrical properties.
Surface resitivity: 4.5×1011 Ohm
Dart impact: 350 g
Static charge decay: 0.5 s, to OV (average)
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 12243-100
- 12243-130
- 12243-074
- 12243-028
- 12243-104
- 12243-134
- 12243-022
- 12243-062
- 12243-032
- 12243-068
- 12243-040
- 12243-064
- 12243-126
- 12243-086
- 12243-120
- 89089-120
- 12243-054
- 12243-052
- 12243-116
- Elongation at break:
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- 90 - 800%
- Melting point:
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- 95...130 °C (203...266 °F)
- Water absorption:
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- <0.01% at 24 h
- Tensile strength:
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- 2700 psi
- Thickness:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil