Chemware® Graduated Cylinders, Teflon® PFA, Saint-Gobain Performance Plastics
Fournisseur: Saint Gobain Performance Plastics
Certificats
A propos de cet article
All the advantages of non-stick, inert Teflon® are available in these graduated cylinders.
Made from the fluoropolymer with the lowest metal ion content, they are excellent for trace analysis. Temperature resistant from the cryogenic range to 500°F. Red markings for easy reading. Smooth pouring spout and stable base.
Spécifications
- Catalog No:
- 24782-053
- 24782-050
- Form:
- Short
- Short