VITLAB® Graduated Cylinders, PMP, Class A, Certified, BrandTech
Fournisseur: Brandtech
Certificats
A propos de cet article
Made of polymethylpentene (PMP) with ring-molded graduations at major increments for excellent readability
- Glass clear PMP
- Certificate supplied
Graduated at increments of 1 or 2% of nominal volume for precise measuring. Cylinders are marked with 'H', lot number, and year of production.
Certifications: Class A per DIN 12681 with test certificate.
Spécifications
- Catalog No:
- 83008-928
- 83008-898
- 10117-442
- 83008-868
- 83008-918
- 83008-888
- 83008-878
- 83008-909
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short