Cylinder Bottles, PVC, Narrow Mouth, Qorpak®
Fournisseur: Qorpak
Certificats
A propos de cet article
Clear PVC Cylinder Bottles provide an excellent resistance to oxygen permeation, oils and good water barrier characteristics.
- Transparent, rigid cylinders allow for easy visual identification of the amount of product inside
Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling.
Customers Who Bought This Also Bought