VWR® Alarm Thermometer/Timer
VWR® Alarm Thermometer/Timer
Alarm Thermometer/Timer, 0...200 °C (32...392 °F)
Catalog # 62344-916
Supplier:   VWR International
VWR® Alarm Thermometer/Timer
Catalog # 62344-916
Supplier:  VWR International
VWR® Alarm Thermometer/Timer
VWR® Alarm Thermometer/Timer
Alarm Thermometer/Timer, 0...200 °C (32...392 °F)
Price
$173.83
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Unit is ideal for monitoring liquids, air/gas, or semi-solids in water baths, ovens, refrigerators, incubators, and tanks.
  • Traceable®
  • Reads °C and °F
  • Easy-to-read display
  • Chemical-resistant ABS base
Easy-to-read “pop-up” display shows time remaining, probe temperature, and temperature alarm setting
Timer counts down from 23 hours, 59 minutes, 59 seconds in one-minute increments. Alarm sounds when time reaches zero. LCD flashes a times-up message and timer begins counting up. For repetitive applications, memory returns time to previous programmed setting.
Fold-back display panel and magnets on the back allow for vertical mounting. Base is made of high-impact, chemical-resistant ABS.
Dual alarms sound separately for programmed time and temperature settings. Five-minute alarm sounds when temperature reading rises above programmed setting. Alarm continues to sound until switched off or temperature returns to an “in-range” condition.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a detachable 21.6 cm (81/2") curved probe with 107 cm (42") long cable and a replaceable AAA alkaline battery.
Services: VWRCATALYST™ provides convenient and efficient re-calibration services for your digital thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. All the service will be performed by an ISO17025 certified lab. Recalibration, certification and shipping fees are all included in the catalog number cost. Simply order the stated catalog number based on how many calibration points are desired. You will receive a protective box to ship your instrument to Avantor's certified calibration laboratory along with a work order form and a prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defects and the performance of your instrument will be compared to certified standards. After re-calibration, your calibration report and instrument will be carefully repacked then returned with standard ground shipping. We also provide expedited service options available upon request.
Specifications
  • Désignation:
    Alarm Thermometer/Timer
  • Range:
    0...200 °C (32...392 °F)
  • Cat. No.:
    62344-916
  • Probe cable length:
    42"
  • No. of channels:
    1
  • W×D×H:
    2.75"×3"×0.5"
  • Temperature range:
    0…200 °C
  • Probe:
    8.5" stainless steel
  • Temperature resolution:
  • Temperature accuracy:
    ±2°C
  • Weight:
    4.25 oz.