VWR® Single Scale Graduated Cylinders, PP
Fournisseur: VWR International
Certificats
A propos de cet article
The “nonwetting” polypropylene (PP) plastic surface of these single-scale graduated cylinders eliminates concave meniscus and makes reading easier and more accurate.
- Polypropylene construction
- Chemically resistant
- Autoclavable
- Single metric scale has raised numerals for easy reading
- Flared top rims and large spouts facilitate pouring
- Octagonal-shaped bases prevent tipping and rolling
Chemically resistant, cylinders are autoclavable at 121°C (250°F).
Spécifications
- Catalog No:
- 65000-000
- 65000-004
- 65000-002
- 65000-008
- 65000-006
- 65000-012
- 65000-010
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short