PYREX® Double Metric Scale Graduated Cylinders, Class A, To Deliver, Corning
Fournisseur: Corning
Certificats
A propos de cet article
PYREX® borosilicate glass, tall form, class A.
- Calibrated to deliver (TD) in accordance with ASTM E-1272 and ASTM E-542
- Double metric scale is numbered both up and down in durable white or blue enamel
- Hexagonal base and pourout
- With shock absorbing polyethylene bumper guards
Spécifications
- Catalog No:
- 89090-490
- 89090-488
- 89090-506
- 89090-504
- 89090-502
- 89090-500
- 89090-498
- 22877-030
- 22877-152
- 22877-150
- 22877-032
- 89090-496
- 89090-494
- 89090-492
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey