PYREX® Single Metric Scale Graduated Cylinders, To Contain, Corning
Supplier: Corning
Certificates
About this item
Borosilicate glass, without stoppers.
- Calibrated 'to contain' (TC)
- Equipped with a 24/40 ST joint
- Single metric scale
- Graduations are in durable white enamel
Specifications
- Catalog No:
- 24763-240
- 24763-266
- 24763-244
- Form:
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey