PYREX® VISTA™ Economy Graduated Cylinders, To Contain, Corning
Fournisseur: Corning
Certificats
A propos de cet article
Made of PYREX® glass, blue graduations.
- Calibrated to contain (TC)
- Tolerances are ±5% of total at any point
- 10 ml cylinder has a funnel top
They are supplied in two parts - a strong, accurate glass graduate and a detachable, sturdy plastic base which absorbs impacts and reduces breakage. Cylinders can be conveniently stacked when plastic bases are removed.
Delivery: Supplied with bumper guards for cylinders of capacity 25 to 100 ml. Snap caps are supplied with all sizes.
Spécifications
- Catalog No:
- 89090-120
- 89090-126
- 89090-114
- 89090-124
- Form:
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey