A propos de cet article
Accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor.
Accurately monitor temperatures in freezers, water baths, heating blocks, incubators, and refrigerators with this enclosed temperature-buffered sensor. Bottle insulates sensor from rapid temperature changes when refrigerator door is opened.
- Bottle insulates sensor from rapid temperature changes
- Solid-state probe eliminates mercury contamination in refrigerators
- Triple display design
Triple display simultaneously shows minimum, maximum and current temperatures.
Triple display simultaneously shows minimum, maximum and current temperatures.
Minimum/maximum monitors high/low readings overnight, on weekends, and for any time period - a significant advantage over current reading-only glass thermometers. Range is -58 to 158°F and -50 to 70°C with a resolution of 0.1° and an accuracy of ± 0.5°C. Alarm feature provides alert when temperature rises above or falls below a set point. Alarm is programmable in 1° increments. Ten-foot micro-cable permits refrigerator doors to close on it.
Probe is sealed in a miniature bottle filled with nontoxic glycol. Solution is GRAS generally recognized as safe by the US FDA.
Spécifications
- Catalog No:
- 89094-782
- 12777-838
- Temperature resolution:
- 0.1°
- 0.1°
- Temperature accuracy:
- ±0.5°C
- ±0.5°C