KIMAX® Graduated Mixing Cylinders, Class B, To Contain, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Calibrated to contain with a durable blue enamel ceramic single metric scale
Cylinders are designed from ASTM Specification E1272, Style 2, Class B requirements.
These mixing cylinders are made of glass.
Cylinders are supplied with a [ST] glass stopper. Height is measured without the stopper.
Specifications
- Catalog No:
- 89001-226
- 89001-228
- 89001-222
- 89001-224
- 89001-218
- 89001-220
- 89000-768
- 89001-230
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall