SP Wilmad-LabGlass Graduated Cylinders, Class B, To Deliver, SP Industries
Fournisseur: SP Industries
Certificats
A propos de cet article
Featuring a reinforced bead and pouring lip, these Class B borosilicate glass graduated cylinders comply with ASTM E 1272
- Reference No. ASTM E 1272
The 5mL size has a funnel top. Calibrated "to deliver" (TD) at 20°C and suitable for general laboratory use.
Spécifications
- Catalog No:
- 14203-718
- 14204-782
- 14208-526
- 14208-376
- Form:
- Short
- Short
- Short
- Short