A propos de cet article
Epredia™ stain kits and replacement components contain all the necessities of the staining procedure.
Gram stain (film) kits identify gram-positive microorganisms with purple and gram-negative bacteria with pink to red. Gram stain (tissue) kits identify gram-positive microorganisms with blue to blue-black and gram-negative microorganisms with red.
- Save time
- Components available separately
- All the necessary stains, solutions and components
Epredia™ stain kits and replacement components contain all the necessities of the staining procedure.
Acid Fast Bacillus, blue secondary stain kits stain bright red with a pale blue background. Acid Fast Bacillus, green secondary stain kit stains dark red with a pale green background. Demonstrate amyloid with Epredia™ Amyloid Stain. Amyloid stain kit stains orange to red.
Acid Fast Bacillus, blue secondary stain kits stain bright red with a pale blue background. Acid Fast Bacillus, green secondary stain kit stains dark red with a pale green background. Demonstrate amyloid with Epredia™ Amyloid Stain. Amyloid stain kit stains orange to red.
Demonstrate the atrophy, thinning and loss of elastic tissue with Epredia™ Elastic Stain. Elastic stain kits stain blue-black to black for elastic tissue, red for collagen with a yellow background.
Save time with Epredia™ Gomori Trichrome (Blue Collagen), a one-step trichrome stain that achieves color differentiation similar to the Masson Trichrome. Gomori Trichrome (Blue Collagen) stain kit stains red for Muscle tissue and cytoplasm, blue for collagen and blue-black for nuclei. Epredia™ Gomori Trichrome (Green Collagen is a one-step trichrome stain that provides distinctive red muscle fibers with green collagen staining.
Detect the presence of ferric iron (hemosiderin) in histological or hematological preparations with Epredia™ Iron Stain. Iron stain kits stain blue iron, pale pink cytoplasm and pink-red nuclei.
Distinguish fungi, basement membranes and opportunistic organisms such as Pneumocystis Carinii in tissue sections with Epredia™ Modified Grocott's Methenamine Silver (Periodic Acid), a uniquely modified silver stain formulation that utilizes methenamine/borax capsules to make working silver solution. Modified Grocott’s Methenamine kit stains black with a green background.
Identify primary tumor sites with the aid of Epredia™ Mucicarmine. Mucicarmine stain kit stains dark rose for mucin and black for nuclei with a yellow background.
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Periodic Acid-Schiff (PAS) Solution. Periodic Acid Schiff kit stains rose to magenta with blue nuclei.
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Steiner-Steiner Silver. Steiner-Steiner silver kits stain black with a yellow to brown background.
Demonstrate the patterns and presence of specific carbohydrates with Epredia™ Periodic Acid-Schiff (PAS) Solution. Periodic Acid Schiff kit stains rose to magenta with blue nuclei.
Determine the presence of spirochetes and nonfilamentous bacteria with Epredia™ Steiner-Steiner Silver. Steiner-Steiner silver kits stain black with a yellow to brown background.