Nalgene® Boston Round Bottles, HDPE, Narrow Mouth, Bulk Packed, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
A propos de cet article
These high-density polyethylene bottles are ideal for storing, packaging, and shipping liquids
- Bottles are available in both translucent and amber formats
- Translucence allows easy viewing of liquid levels
- Amber reduces UV light transmissions to protect light-sensitive liquids
They offer excellent chemical resistance, and are highly reliable and durable for long-term use. Leakproof.
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Bulk packed for packaging applications. Bottles and closures are packed in separate polybags within the same carton.
Customers Who Bought This Also Bought