Wood Funnel Support, Adjustable Bracket, Bennett Wood
# de catalogue 30575-001
Fournisseur: Bennett Wood Specialties
Spécifications
- Description:Funnel Support
- Cat. no.:30575-001
- Supplier No.:2608
Spécifications
A propos de cet article
Holds six funnels
- Without glassware
- With bracket adjustable on each end
Shelf is adjustable over a height of 127 mm (5"). Slotted funnel openings spaced 133 mm (51/4") center to center.
Customers Who Bought This Also Bought