Wood Funnel Support, Bennett Wood
# de catalogue 30580-001
Fournisseur: Bennett Wood Specialties
Spécifications
- Description:Funnel Support
- Cat. no.:30580-001
- Supplier No.:2635
Spécifications
A propos de cet article
Designed to hold six separatory funnels
With adjustable upper arm and auxiliary middle arm similarly slotted, and adjustable in height. Without glassware.
Customers Who Bought This Also Bought