Nunc® CryoFlex™ Tubing, Thermo Scientific
# de catalogue CA24270-013
Fournisseur: Thermo Fisher Scientific
Spécifications
- Length:500 mm (1911/16")
- Description:CryoFlex™ tubing, 14 mm diameter
- Cat. no.:CA24270-013
- Supplier No.:343958
Spécifications
A propos de cet article
Flexible polyethylene tubing provides extra safety when freezing hazardous samples or storing in liquid nitrogen
Can be used as a cane to lower CryoTube™ vials into liquid nitrogen.
Customers Who Bought This Also Bought