Nalgene® Boston Round Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Ideal for lab applications.
- Excellent chemical resistance
- With polypropylene screw closures
- Leakproof, Autoclavable
Note: Before autoclaving, set closure on top of bottle without engaging threads.
The 3.8 and 8 L sizes have a built-in shoulder loop for attaching an identification tag.
Ordering information: For replacement closures, see 16219-122 series.
Specifications
- Catalog No:
- 16067-102
- 16066-960
- 16067-124
- 16067-022
- 16067-044
- 16066-981
- 16067-000
- 16067-066
- 16067-088
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO14001 (Certificate Registration No. EMS 752603)
- ISO14001 (Certificate Registration No. EMS 752603)