KIMAX® Filling Funnels, Long Stem, 58° Angle, Kimble Chase, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Constructed with a strong, molded rim, heavy uniform walls, and a fire-polished stem for a long service life
Unique 58° funnel angle provides faster and more effective filtration than the standard 60° funnel design. Funnels also have a high resistance to chemical attack and mechanical and thermal shock.
Certifications: Designed from ASTM Specification E1095, Type 1, Class B requirements.
Spécifications
- Catalog No:
- 89000-824
- 89001-400
- 89001-398
- Stem length:
- 150 mm
- 150 mm
- 150 mm